Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2-Piperazinecarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A425161-100G A425161-10G A425161-500G A425161-1G A425161-25G A425161-5G
103827-458EA 5 USD InStock
103827-458 103838-508 76829-460 76829-462 76829-464 76842-158 76842-160 76842-162
(±)-2-Piperazinecarboxylic acid 97%
(±)-2-Piperazinecarboxylic acid
Precaución: for Research Use Only


Formula: C₅H₁₀N₂O₂
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00132878
CAS Number: 2762-32-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty