Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium [(benzyloxy)methyl]trifluoroborate 95%

Supplier: AMBEED, INC
Warning

A160227-1G A160227-5G A160227-10G A160227-250MG A160227-1G A160227-5G
103805-352EA 26 USD InStock
103805-352 103843-582 76973-540 76973-542 77300-452 77300-454
Potassium [(benzyloxy)methyl]trifluoroborate 95%
Potassium [(benzyloxy)methyl]trifluoroborate
Precaución: for Research Use Only


Formula: C₈H₉BF₃O·K
MW: 228.061 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11052815
CAS Number: 1027642-25-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty