Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium-4-tert-butylphenyltrifluoroborate 98%

Supplier: AMBEED, INC
Danger

A302825-1G A302825-10G A302825-1G A302825-5G
103814-350EA 6 USD InStock
103814-350 103835-510 76723-636 76838-196 76838-198
Potassium-4-tert-butylphenyltrifluoroborate 98%
Potassium 4-tert-butylphenyltrifluoroborate
Precaución: for Research Use Only


Formula: C₁₀H₁₃BF₃K
MW: 240.11 g/mol
Melting Pt: >230 °C
Storage Temperature: Ambient
MDL Number: MFCD06659940
CAS Number: 423118-47-2
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty