Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium bromo(difluoro)acetate 95%

Supplier: AMBEED, INC
Warning

A156170-1G A156170-5G A156170-100G A156170-1G A156170-25G
103804-842EA 9 USD InStock
103804-842 103843-046 77162-954 77318-796 77318-798
Potassium bromo(difluoro)acetate 95%
Potassium bromo(difluoro)acetate
Precaución: Keep in a dark place, inert atmosphere, room temperature


Formula: C₂BrF₂O₂·K
MW: 213.019 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784243
CAS Number: 87189-16-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty