Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium cyclohexyltrifluoroborate 96%

Supplier: AMBEED, INC
Danger

A506977-1G A506977-5G A506977-100MG A506977-250MG
103828-338EA 10 USD InStock
103828-338 103839-440 76843-420 76843-422 76878-086 76878-088
Potassium cyclohexyltrifluoroborate 96%
Potassium cyclohexyltrifluoroborate
Precaución: for Research Use Only


Formula: C₆H₁₁BF₃K
Melting Pt: 187…192 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD09992929
CAS Number: 446065-11-8
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty