Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium cyclopentyltrifluoroborate 97%

Supplier: AMBEED, INC
Warning

A134994-1G A134994-100MG A134994-10G A134994-250MG A134994-1G A134994-25G A134994-5G
103802-080EA 8 USD InStock
103802-080 76870-950 76870-952 76870-954 77314-856 77314-858 77314-860
Potassium cyclopentyltrifluoroborate 97%
Potassium cyclopentyltrifluoroborate
Precaución: for Research Use Only


Formula: C₅H₉BF₃K
Storage Temperature: Freezer
MDL Number: MFCD04112712
CAS Number: 1040745-70-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty