Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium cyclopropyltrifluoroborate 97%

Supplier: AMBEED, INC
Warning

A172269-1G A172269-100G A172269-10G A172269-250MG A172269-500G A172269-1G A172269-25G A172269-5G
103806-708EA 6 USD InStock
103806-708 103844-992 76813-232 76813-234 76813-236 76813-238 77082-178 77082-180 77082-182
Potassium cyclopropyltrifluoroborate 97%
Potassium cyclopropyltrifluoroborate
Precaución: for Research Use Only


Formula: C₃H₅BF₃K
Melting Pt: 348…350 °C
Storage Temperature: Ambient
MDL Number: MFCD09265031
CAS Number: 1065010-87-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty