Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tricarballylic acid 97%

Supplier: AMBEED, INC
Warning

A263335-5G A263335-100G A263335-10G A263335-25G A263335-5G
103852-504EA 15 USD InStock
103852-504 76787-034 76787-036 77092-142 77304-868
Tricarballylic acid 97%
Tricarballylic acid
Precaución: for Research Use Only


Formula: C₆H₈O₆
MW: 176.13 g/mol
Boiling Pt: 266
Melting Pt: 160
Storage Temperature: Ambient
MDL Number: MFCD00002723
CAS Number: 99-14-9
Merck Index: 12,09752
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty