Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Propylhydrazine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A412383-10G A412383-250MG A412383-1G A412383-5G
103827-248EA 55 USD InStock
103827-248 103838-294 76817-982 76817-984 76841-836 76841-838
1-Propylhydrazine hydrochloride 95%
1-Propylhydrazine hydrochloride


Formula: C₃H₁₀N₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD01320515
CAS Number: 56795-66-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty