Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxypyrazine 95%

Supplier: AMBEED, INC
Warning

A125973-1G A125973-100G A125973-10G A125973-1G A125973-5G
103800-744EA 16 USD InStock
103800-744 77258-274 77258-276 77312-908 77312-910
2-Hydroxypyrazine 95%
2-Hydroxypyrazine
Precaución: for Research Use Only


Formula: C₄H₄N₂O
Melting Pt: 185…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00233966
CAS Number: 6270-63-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty