Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrazine-2,6-dicarboxylic acid 95%

Supplier: AMBEED, INC
Warning

A132260-1G A132260-100MG A132260-250MG A132260-1G
103801-706EA 82 USD InStock
103801-706 76811-272 76811-274 77314-276
Pyrazine-2,6-dicarboxylic acid 95%
Pyrazine-2,6-dicarboxylic acid
Precaución: for Research Use Only


Formula: C₆H₄N₂O₄
MW: 168.11 g/mol
Melting Pt: 225 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08694102
CAS Number: 940-07-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty