Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrazine-2-carbohydrazide 97%

Supplier: AMBEED, INC
Warning

A212601-1G A212601-5G A212601-1G A212601-250MG A212601-25G
103818-216EA 13 USD InStock
103818-216 103848-912 77087-378 77164-996 77164-998
Pyrazine-2-carbohydrazide 97%
Pyrazine-2-carbohydrazide
Precaución: for Research Use Only


Formula: C₅H₆N₄O
Melting Pt: 169 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00778762
CAS Number: 768-05-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty