Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrazolo[1,5-a][1,3,5]triazin-4(3H)-one 97%

Supplier: AMBEED, INC
Warning

A103020-1G A103020-5G A103020-10G A103020-250MG A103020-1G A103020-5G
103796-468EA 70 USD InStock
103796-468 103822-170 76869-216 76869-218 77309-036 77309-038
Pyrazolo[1,5-a][1,3,5]triazin-4(3H)-one 97%
Pyrazolo[1,5-a][1,3,5]triazin-4(3H)-one
Precaución: for Research Use Only


Formula: C₅H₄N₄O
Flash Pt: 95.7 °C
Storage Temperature: Ambient
MDL Number: MFCD20617746
CAS Number: 54346-27-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty