Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridazine 98%

Supplier: AMBEED, INC
Warning

A148352-1G A148352-5G A148352-100G A148352-10G A148352-25G A148352-1G A148352-5G
103803-922EA 11 USD InStock
103803-922 103842-034 76719-632 76719-634 76719-636 77317-492 77317-494
Pyridazine 98%
Pyridazine
Precaución: for Research Use Only


Formula: C₄H₄N₂
MW: 80.09 g/mol
Boiling Pt: 208
Melting Pt: 0
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00006463
CAS Number: 289-80-5
Merck Index: 12,08152
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty