Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridine-3-sulfonyl chloride 98%

Supplier: AMBEED, INC
Danger

A626684-5G A626684-1G A626684-25G A626684-5G A626684-10G
103840-256EA 7 USD InStock
103840-256 76844-422 76844-424 76844-426 76980-656
Pyridine-3-sulfonyl chloride 98%
Pyridine-3-sulfonyl chloride
Precaución: For research use only


Formula: C5H4ClNO2S
MW: 177.6088 g/mol
Flash Pt: 125.657 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03452747
CAS Number: 16133-25-8
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty