Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrido[3,2-d]pyrimidin-4(3H)-one 98%

Supplier: AMBEED, INC
Warning

A190419-1G A190419-5G A190419-100MG A190419-250MG
103816-294EA 54 USD InStock
103816-294 103846-894 76825-252 76825-254
Pyrido[3,2-d]pyrimidin-4(3H)-one 98%
Pyrido[3,2-d]pyrimidin-4(3H)-one
Precaución: for Research Use Only


Formula: C₇H₅N₃O
MW: 147.134 g/mol
Boiling Pt: 346.2 °C (760 mm Hg)
Flash Pt: 163 °C
Storage Temperature: Ambient
MDL Number: MFCD02645992
CAS Number: 37538-67-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty