Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrimidine-4-carbaldehyde 95%

Supplier: AMBEED, INC
Warning

A357529-5G A357529-1G A357529-250MG
103826-196EA 168 USD InStock
103826-196 103837-200 76840-356 77115-234
Pyrimidine-4-carbaldehyde 95%
Pyrimidine-4-carbaldehyde
Precaución: for Research Use Only


Formula: C₅H₄N₂O
MW: 108.1 g/mol
Boiling Pt: 225.3 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD03789624
CAS Number: 2435-50-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty