Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Quinazolinecarboxylic acid 96%

Supplier: AMBEED, INC
Warning

A218421-1G A218421-250MG A218421-1G
103818-712EA 444 USD InStock
103818-712 76785-890 77303-490
6-Quinazolinecarboxylic acid 96%
6-Quinazolinecarboxylic acid


Formula: C₉H₆N₂O₂
Boiling Pt: 359.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11520386
CAS Number: 676326-53-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty