Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Quinolineboronic acid 97%

Supplier: AMBEED, INC
Warning

A273196-1G A273196-5G A273196-5G A273196-10G A273196-250MG A273196-25G
103813-134EA 190 USD InStock
103813-134 103853-054 77092-852 77262-524 77262-526 77305-126
5-Quinolineboronic acid 97%
5-Quinolineboronic acid
Precaución: for Research Use Only


Formula: C₉H₈BNO₂
MW: 172.98 g/mol
Boiling Pt: 400.3 °C (760 mm Hg)
Melting Pt: 144…148 °C
Flash Pt: 195.9 °C
Storage Temperature: Ambient
MDL Number: MFCD03095058
CAS Number: 355386-94-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty