Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Quinoxalinecarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A108074-1G A108074-5G A108074-10G A108074-250MG A108074-1G
103797-288EA 29 USD InStock
103797-288 103823-062 77160-574 77160-576 77309-906
5-Quinoxalinecarboxylic acid 97%
5-Quinoxalinecarboxylic acid
Precaución: for Research Use Only


Formula: C₉H₆N₂O₂
Boiling Pt: 394.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD02854494
CAS Number: 6924-66-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty