Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ro 15-3890 98%

Supplier: AMBEED, INC
Warning

A112716-5MG A112716-25MG A112716-50MG A112716-1MG A112716-25MG A112716-50MG A112716-5MG A112716-100MG
103791-424EA 45 USD InStock
103791-424 103792-026 103799-282 77284-260 77310-736 77310-738 77310-740 77747-824
Ro 15-3890 98%
Ro 15-3890
Precaución: for Research Use Only


Formula: C₁₃H₁₀FN₃O₃
Boiling Pt: 570.5 °C (760 mm Hg)
Flash Pt: 298.8 °C
Storage Temperature: Freezer
MDL Number: MFCD08061420
CAS Number: 84378-44-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty