Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rofecoxib 98%

Supplier: AMBEED, INC
Warning

A395001-1G A395001-100MG A395001-1MG A395001-250MG A395001-25MG
103826-956EA 61 USD InStock
103826-956 77115-674 77115-676 77115-678 77115-680
Rofecoxib 98%
Rofecoxib
Precaución: for Research Use Only


Formula: C₁₇H₁₄O₄S
Boiling Pt: 577.6 °C (760 mm Hg)
Melting Pt: 207 °C
Flash Pt: 303.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00935806
CAS Number: 162011-90-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty