Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rosiglitazone 98%

Supplier: AMBEED, INC
Warning

A158185-1G A158185-5G A158185-1MG A158185-25G A158185-50MG A158185-1G
103805-102EA 32 USD InStock
103805-102 103843-310 76997-654 76997-656 76997-658 77319-162
Rosiglitazone 98%
Rosiglitazone
Precaución: for Research Use Only


Formula: C₁₈H₁₉N₃O₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00871760
CAS Number: 122320-73-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty