Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ruthenium(III) chloride 97%

Supplier: AMBEED, INC
Danger

A275839-1G A275839-5G A275839-10G A275839-1G A275839-25G A275839-5G
103813-280EA 28 USD InStock
103813-280 103853-206 76723-142 77305-214 77305-216 77305-218
Ruthenium(III) chloride 97%
Ruthenium(III) chloride
Precaución: for Research Use Only


Formula: Cl₃Ru
MW: 207.43
Melting Pt: 500
Density: 3.11
Storage Temperature: Ambient
MDL Number: MFCD00011208
CAS Number: 10049-08-8
UN: 3260
ADR: 8 Ⅱ
Merck Index: 12,08455
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty