Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

S-(Benzo[d]thiazol-2-yl)-N,N-dicyclohexylthiohydroxylamine 95%

Supplier: AMBEED, INC
Warning

A153665-1G A153665-5G A153665-100G A153665-10G A153665-250MG A153665-500G A153665-1G A153665-25G A153665-5G
103804-548EA 7 USD InStock
103804-548 103842-722 76973-210 76973-212 76973-214 76973-216 77318-394 77318-396 77318-398
S-(Benzo[d]thiazol-2-yl)-N,N-dicyclohexylthiohydroxylamine 95%
S-(Benzo[d]thiazol-2-yl)-N,N-dicyclohexylthiohydroxylamine
Precaución: for Research Use Only


Formula: C₁₉H₂₆N₂S₂
Boiling Pt: 486.5 °C (760 mm Hg)
Melting Pt: 104 °C
Flash Pt: 248 °C
Storage Temperature: Ambient
MDL Number: MFCD00236063
CAS Number: 4979-32-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty