Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-2-Amino-3-methyl-1,1-diphenyl-1-butanol 97%

Supplier: AMBEED, INC
Warning

A100324-1G A100324-5G A100324-100MG A100324-10G A100324-250MG A100324-25G A100324-1G A100324-5G
103795-368EA 21 USD InStock
103795-368 103821-070 77135-426 77135-428 77135-430 77135-432 77296-636 77296-638
(S)-(-)-2-Amino-3-methyl-1,1-diphenyl-1-butanol 97%
(S)-(-)-2-Amino-3-methyl-1,1-diphenyl-1-butanol
Precaución: for Research Use Only


Formula: C₁₇H₂₁NO
Melting Pt: 95…99 °C
Storage Temperature: Ambient
MDL Number: MFCD01318248
CAS Number: 78603-95-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty