Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sarafloxacin hydrochloride 98%

Supplier: AMBEED, INC
Warning

A129788-1G A129788-100G A129788-100MG A129788-1MG A129788-500G A129788-1G A129788-5G A129788-25G
103801-324EA 21 USD InStock
103801-324 76767-142 76767-144 76767-146 76767-148 77298-370 77313-712 77876-936
Sarafloxacin hydrochloride 98%
Sarafloxacin hydrochloride
Precaución: for Research Use Only


Formula: C₂₀H₁₇F₂N₃O₃·HCl
Melting Pt: >275 °C
Storage Temperature: Ambient
MDL Number: MFCD03427297
CAS Number: 91296-87-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty