Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(-)-Shikimic acid 98%

Supplier: AMBEED, INC
Warning

A134982-100G A134982-500G A134982-25G A134982-1000G
76767-406EA 27 USD InStock
76767-406 76767-408 76767-410 76767-412 76767-414 77314-850 77624-132
(-)-Shikimic acid 98%
(-)-Shikimic acid
Precaución: for Research Use Only


Formula: C₇H₁₀O₅
MW: 174.15 g/mol
Boiling Pt: 400.5 °C (760 mm Hg)
Melting Pt: 185…187 °C
Storage Temperature: Freezer
MDL Number: MFCD00066278
CAS Number: 138-59-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty