Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium (R)-3-hydroxybutanoate 97%

Supplier: AMBEED, INC
Warning

A288555-1G A288555-1G A288555-5G A288555-10G A288555-250MG A288555-25G A288555-500G
103813-802EA 8 USD InStock
103813-802 103834-944 76837-508 76837-510 77141-300 77141-302 77305-534 77746-534
Sodium (R)-3-hydroxybutanoate 97%
Sodium (R)-3-hydroxybutanoate
Precaución: for Research Use Only


Formula: C₄H₇NaO₃
Boiling Pt: 269.2 °C (760 mm Hg)
Melting Pt: 149…155 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077788
CAS Number: 13613-65-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty