Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-1-methyl-3-sulfinopropionate 95%

Supplier: AMBEED, INC
Warning

A109331-1G A109331-5G A109331-100MG A109331-10G A109331-250MG A109331-5G
103797-494EA 30 USD InStock
103797-494 103823-290 76717-998 76718-000 76718-002 77283-862
Sodium-1-methyl-3-sulfinopropionate 95%
Sodium-1-methyl-3-sulfinopropionate
Precaución: For research use only.


Formula: C₄H₈O₄S
Melting Pt: 199…216 °C
Storage Temperature: Ambient
MDL Number: MFCD07776796
CAS Number: 90030-48-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty