Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 4-nitrobenzoate 95%

Supplier: AMBEED, INC
Warning

A227222-1G A227222-250MG A227222-5G A227222-1G
103819-308EA 8 USD InStock
103819-308 76975-988 76975-990 77303-796
Sodium 4-nitrobenzoate 95%
Sodium 4-nitrobenzoate
Precaución: for Research Use Only


Formula: C₇H₄NNaO₄
Boiling Pt: 359.1 °C (760 mm Hg)
Flash Pt: 166.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00797416
CAS Number: 3847-57-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty