Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium pivalate hydrate 95%

Supplier: AMBEED, INC
Warning

A240989-5G A240989-100G A240989-10G A240989-500G A240989-25G A240989-5G
103851-190EA 5 USD InStock
103851-190 76722-440 76722-442 76722-444 77090-346 77090-348
Sodium pivalate hydrate 95%
Sodium pivalate hydrate
Precaución: for Research Use Only


Formula: C₅H₁₁NaO₃
MW: 142.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150782
CAS Number: 143174-36-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty