Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sulfaphenazole 98%

Supplier: AMBEED, INC
Warning

A266126-25MG A266126-1G A266126-1MG A266126-250MG
103792-522EA 20 USD InStock
103792-522 76787-086 76787-088 77668-146
Sulfaphenazole 98%
Sulfaphenazole
Precaución: for Research Use Only


Formula: C₁₅H₁₄N₄O₂S
Boiling Pt: 541.9 °C (760 mm Hg)
Melting Pt: 179…183 °C
Flash Pt: 281.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00057226
CAS Number: 526-08-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty