Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Swertiamarin 97%

Supplier: AMBEED, INC
Warning

A513464-10MG A513464-1MG A513464-5MG A513464-100MG A513464-250MG A513464-1G
103799-670EA 52 USD InStock
103799-670 76790-024 76790-026 76790-028 77669-278 77669-280 77872-855
Swertiamarin 97%
Swertiamarin
Precaución: for Research Use Only


Formula: C₁₆H₂₂O₁₀
Boiling Pt: 649.3 °C (760 mm Hg)
Melting Pt: 111 °C
Flash Pt: 237.7 °C
Storage Temperature: Freezer
MDL Number: MFCD07783984
CAS Number: 17388-39-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty