Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tazobactam ≥98%

Supplier: AMBEED, INC
Danger

A289987-250MG A289987-10G
103813-856EA 39 USD InStock
103813-856 103834-996 77616-258 77685-829
Tazobactam ≥98%
Tazobactam
Precaución: Sealed in dry, store in freezer, under –20 °C.


Formula: C₁₀H₁₂N₄O₅S
Boiling Pt: 707.1 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD00867002
CAS Number: 89786-04-9
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty