Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl (2-hydroxyethyl)(methyl)carbamate 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A232744-5G A232744-25G A232744-5G A232744-10G A232744-100G A232744-500G
103850-600EA 5 USD InStock
103850-600 77089-532 77089-534 77165-574 77568-256 77683-680
tert-Butyl (2-hydroxyethyl)(methyl)carbamate 97%, Ambeed.Inc
tert-Butyl (2-hydroxyethyl)(methyl)carbamate
Precaución: For research use only.


Formula: C₈H₁₇NO₃
Boiling Pt: 245.1 °C (760 mm Hg)
Flash Pt: 102.1 °C
Storage Temperature: Freezer
MDL Number: MFCD03425857
CAS Number: 57561-39-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty