Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-3-hydroxycyclobutylcarbamate 97%

Supplier: AMBEED, INC
Warning

A171653-1G A171653-5G A171653-100G A171653-10G A171653-250MG A171653-1G A171653-25G A171653-5G
103806-652EA 10 USD InStock
103806-652 103844-928 77029-046 77029-048 77029-050 77082-090 77082-092 77082-094
tert-Butyl-3-hydroxycyclobutylcarbamate 97%
tert-Butyl-3-hydroxycyclobutylcarbamate
Precaución: for Research Use Only


Formula: C₉H₁₇NO₃
MW: 187.24 g/mol
Boiling Pt: 303.7 °C (760 mm Hg)
Flash Pt: 137.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11501030
CAS Number: 154748-63-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty