Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate 98%

Supplier: AMBEED, INC
Warning

A155552-1G A155552-5G A155552-100MG A155552-250MG A155552-5G A155552-1G
103804-778EA 73 USD InStock
103804-778 103842-974 76812-452 76812-454 77300-126 77318-700
tert-Butyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate 98%
tert-Butyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate
Precaución: for Research Use Only


Formula: C₁₆H₂₅BN₂O₄
Storage Temperature: Freezer
MDL Number: MFCD12032217
CAS Number: 1095708-32-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty