Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Boc-amino)benzyl alcohol 97%

Supplier: AMBEED, INC
Warning

A194586-5G A194586-100G A194586-10G A194586-250MG A194586-25G A194586-1G A194586-5G
103816-670EA 6 USD InStock
103816-670 103847-280 76691-022 76691-024 76691-026 76691-028 77085-206 77085-208
4-(Boc-amino)benzyl alcohol 97%
4-(Boc-amino)benzyl alcohol
Precaución: for Research Use Only


Formula: C₁₂H₁₇NO₃
MW: 223.268 g/mol
Boiling Pt: 310.049 °C (760 mm Hg)
Flash Pt: 141.313 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04974271
CAS Number: 144072-29-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty