Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-4-bromobutylcarbamate 95%

Supplier: AMBEED, INC
Warning

A123263-1G A123263-250MG A123263-1G A123263-25G
103800-374EA 21 USD InStock
103800-374 77284-956 77312-492 77747-891
tert-Butyl-4-bromobutylcarbamate 95%
tert-Butyl-4-bromobutylcarbamate
Precaución: for Research Use Only


Formula: C₉H₁₈BrNO₂
MW: 252.16 g/mol
Boiling Pt: 308.8 °C (760 mm Hg)
Flash Pt: 140.5 °C
Storage Temperature: Freezer
MDL Number: MFCD06201020
CAS Number: 164365-88-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty