Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl (4-iodocyclohexyl)carbamate 97%

Supplier: AMBEED, INC
Warning

A112525-1G A112525-100MG A112525-250MG A112525-1G
103798-030EA 98 USD InStock
103798-030 77160-816 77160-818 77310-676
tert-Butyl (4-iodocyclohexyl)carbamate 97%
tert-Butyl (4-iodocyclohexyl)carbamate
Precaución: for Research Use Only


Formula: C₁₁H₂₀INO₂
MW: 325.19 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18252818
CAS Number: 1179986-79-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty