Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine 97%

Supplier: AMBEED, INC
Warning

A154972-1G A154972-5G A154972-100MG A154972-250MG
103804-700EA 162 USD InStock
103804-700 103842-896 76997-484 77671-186
cis-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine 97%
cis-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine
Precaución: for Research Use Only


Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Boiling Pt: 322.1±31.0 °C (760 mm Hg)
Flash Pt: 148.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07367780
CAS Number: 184954-75-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty