Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-tert-Butyl-3-hydroxycyclobutylcarbamate 97%

Supplier: AMBEED, INC
Warning

A107090-1G A107090-5G A107090-100G A107090-10G A107090-250MG A107090-5G A107090-1G
103797-126EA 15 USD InStock
103797-126 103822-884 77160-538 77160-540 77160-542 77283-538 77309-754
cis-tert-Butyl-3-hydroxycyclobutylcarbamate 97%
cis-tert-Butyl-3-hydroxycyclobutylcarbamate
Precaución: for Research Use Only


Formula: C₉H₁₇NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09038208
CAS Number: 389890-43-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty