Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl (cis-4-aminocyclohexyl)carbamate 95%

Supplier: AMBEED, INC
Warning

A110989-1G A110989-100G A110989-10G A110989-250MG A110989-25G A110989-1G A110989-5G
103797-772EA 23 USD InStock
103797-772 103823-578 76995-202 76995-204 76995-206 76995-208 77310-386 77310-388
tert-Butyl (cis-4-aminocyclohexyl)carbamate 95%
tert-Butyl (cis-4-aminocyclohexyl)carbamate
Precaución: for Research Use Only


Formula: C₁₁H₂₂N₂O₂
Boiling Pt: 322.1 °C (760 mm Hg)
Melting Pt: 90…94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03844604
CAS Number: 247570-24-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty