Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(N-Boc-methylamino)acetonitrile 97%

Supplier: AMBEED, INC
Warning

A368607-1G A368607-5G A368607-10G A368607-250MG
103826-436EA 34 USD InStock
103826-436 103837-450 77015-946 77015-948
2-(N-Boc-methylamino)acetonitrile 97%
2-(N-Boc-methylamino)acetonitrile
Precaución: for Research Use Only


Formula: C₈H₁₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06658361
CAS Number: 180976-09-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty