Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,8-Diazaspiro[4.5]decane-8-carboxylic acid tert-butyl ester 95%

Supplier: AMBEED, INC
Warning

A183840-1G A183840-5G A183840-100MG A183840-250MG A183840-5G
103815-676EA 119 USD InStock
103815-676 103846-238 76813-668 76813-670 77083-842
1,8-Diazaspiro[4.5]decane-8-carboxylic acid tert-butyl ester 95%
1,8-Diazaspiro[4.5]decane-8-carboxylic acid tert-butyl ester
Precaución: for Research Use Only


Formula: C₁₃H₂₄N₂O₂
Boiling Pt: 337.0±35.0 °C (760 mm Hg)
Flash Pt: 157.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11501187
CAS Number: 937729-06-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty