Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 14-hydroxy-3,6,9,12-tetraoxatetradecan-1-oate 95%

Supplier: AMBEED, INC
Warning

A114623-1G A114623-100MG A114623-250MG A114623-1G A114623-5G
103798-434EA 71 USD InStock
103798-434 77257-842 77257-844 77311-098 77747-831
tert-Butyl 14-hydroxy-3,6,9,12-tetraoxatetradecan-1-oate 95%
tert-Butyl 14-hydroxy-3,6,9,12-tetraoxatetradecan-1-oate
Precaución: for Research Use Only


Formula: C₁₄H₂₈O₇
Storage Temperature: Refrigerator
MDL Number: MFCD27635170
CAS Number: 169751-72-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty