Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl (diethylphosphono)acetate 98%

Supplier: AMBEED, INC
Warning

A153165-1000G A153165-100G A153165-10G A153165-500G A153165-25G
103842-638EA 7 USD InStock
103842-638 76783-456 76783-458 76783-460 76783-462 77318-274
tert-Butyl (diethylphosphono)acetate 98%
tert-Butyl (diethylphosphono)acetate
Precaución: for Research Use Only


Formula: C₁₀H₂₁O₅P
MW: 252.25 g/mol
Boiling Pt: 100…103 °C (1.5 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00075414
CAS Number: 27784-76-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty