Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 2-(hydroxymethyl)-5,6-dihydroimidazo[1,2-a]pyrazine-7(8H)-carboxylate 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A100090-1G A100090-100MG A100090-250MG
103795-244EA 396 USD InStock
103795-244 77160-144 77682-926
tert-Butyl 2-(hydroxymethyl)-5,6-dihydroimidazo[1,2-a]pyrazine-7(8H)-carboxylate 95%, Ambeed.Inc
tert-Butyl 2-(hydroxymethyl)-5,6-dihydroimidazo[1,2-a]pyrazine-7(8H)-carboxylate
Precaución: For research use only.


Formula: C₁₂H₁₉N₃O₃
Boiling Pt: 462.9 °C (760 mm Hg)
Flash Pt: 233.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18792756
CAS Number: 1250996-70-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty