Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(tert-Butoxycarbonylmethylene)triphenylphosphorane 97%

Supplier: AMBEED, INC
Warning

A173483-1G A173483-5G A173483-100G A173483-10G A173483-25G A173483-500G A173483-1G
103806-818EA 5 USD InStock
103806-818 103845-118 77111-112 77111-114 77111-116 77111-118 77301-272
(tert-Butoxycarbonylmethylene)triphenylphosphorane 97%
(tert-Butoxycarbonylmethylene)triphenylphosphorane
Precaución: for Research Use Only


Formula: C₂₄H₂₅O₂P
MW: 376.436 g/mol
Boiling Pt: 503.5 °C (760 mm Hg)
Melting Pt: 146…151 °C
Flash Pt: 271.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075545
CAS Number: 35000-38-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty